missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Chromosorb™ LC-9

Catalog No. 111018015
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018015 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018015 Supplier MilliporeSigma™ Supelco™ Supplier No. C626910G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prepared by chemically bonding an amino type functional group on the microparticle silica substrate.

Specifications

Particle Size 10 μm
Quantity 10 g
Surface Area (Metric) >350 m2/g
Product Line Chromosorb
MDL Number MFCD00166544
Density 0.48 g/cm3 at 25°C (lit.)
CAS 79029-81-5

Chromosorb is a registered trademark of Imerys Minerals California, Inc.