missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Chromosorb™ LC-7

Catalog No. 111012164
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012164 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012164 Supplier MilliporeSigma™ Supelco™ Supplier No. C626710G

A reverse phase microparticulate packing used for separating a variety of compounds used in pharmaceutical and food products.

Specifications

Particle Size 10 μm
Packaging Poly Bottle
Quantity 10 g
Surface Area (Metric) >250 m2/g
Product Line Chromosorb
MDL Number MFCD00166702
Density 0.45 g/cm3 (lit.)
CAS 102634-18-4

Chromosorb is a registered trademark of Imerys Minerals California, Inc.