missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromone-3-carboxaldehyde 98.0+%, TCI America™
$40.43 - $119.36
Chemical Identifiers
| CAS | 17422-74-1 |
|---|---|
| Molecular Formula | C10H6O3 |
| Molecular Weight (g/mol) | 174.155 |
| MDL Number | MFCD00014667 |
| InChI Key | FSMYWBQIMDSGQP-UHFFFAOYSA-N |
| Synonym | chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde |
| PubChem CID | 87112 |
| IUPAC Name | 4-oxochromene-3-carbaldehyde |
| SMILES | C1=CC=C2C(=C1)C(=O)C(=CO2)C=O |
Chemical Identifiers
| 17422-74-1 | |
| 174.155 | |
| FSMYWBQIMDSGQP-UHFFFAOYSA-N | |
| 87112 | |
| C1=CC=C2C(=C1)C(=O)C(=CO2)C=O |
| C10H6O3 | |
| MFCD00014667 | |
| chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde | |
| 4-oxochromene-3-carbaldehyde |
Specifications
| 17422-74-1 | |
| White-Yellow | |
| MFCD00014667 | |
| chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde | |
| C1=CC=C2C(=C1)C(=O)C(=CO2)C=O | |
| 174.155 | |
| 174.16 | |
| Crystalline Powder |
| 153°C | |
| C10H6O3 | |
| 5 g | |
| FSMYWBQIMDSGQP-UHFFFAOYSA-N | |
| 4-oxochromene-3-carbaldehyde | |
| 87112 | |
| ≥98.0% (GC) | |
| Chromone-3-carboxaldehyde |
Safety and Handling
TSCA : No