missing translation for 'onlineSavingsMsg'
Learn More

Chromone-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. O02915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02915G 5 g
O029125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02915G Supplier TCI America Supplier No. O02915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17422-74-1
Molecular Formula C10H6O3
Molecular Weight (g/mol) 174.155
MDL Number MFCD00014667
InChI Key FSMYWBQIMDSGQP-UHFFFAOYSA-N
Synonym chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde
PubChem CID 87112
IUPAC Name 4-oxochromene-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)C=O

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Chromone-3-carboxaldehyde