Learn More
Chromone-3-carboxaldehyde, 97%
CAS: 17422-74-1 | C10H6O3 | 174.155 g/mol
Supplier: Thermo Scientific Chemicals A1543814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Chromone-3-carboxaldehyde | |
| 150°C to 154°C | |
| MFCD00014667 | |
| 472849 | |
| chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde | |
| C1=CC=C2C(=C1)C(=O)C(=CO2)C=O | |
| 174.155 | |
| 174.16 |
| 17422-74-1 | |
| C10H6O3 | |
| 25 g | |
| Air sensitive | |
| FSMYWBQIMDSGQP-UHFFFAOYSA-N | |
| 4-oxochromene-3-carbaldehyde | |
| 87112 | |
| 97% |
Chemical Identifiers
| 17422-74-1 | |
| 174.155 | |
| FSMYWBQIMDSGQP-UHFFFAOYSA-N | |
| 87112 | |
| C1=CC=C2C(=C1)C(=O)C(=CO2)C=O |
| C10H6O3 | |
| MFCD00014667 | |
| chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde | |
| 4-oxochromene-3-carbaldehyde |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 241-451-5
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only