missing translation for 'onlineSavingsMsg'
Learn More

Chromone-3-carboxaldehyde, 97%

Catalog No. AAA1543806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1543806 5 g
AAA1543814 25 g
AAA1543822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1543806 Supplier Thermo Scientific Chemicals Supplier No. A1543806

CAS: 17422-74-1 | C10H6O3 | 174.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17422-74-1
Molecular Formula C10H6O3
Molecular Weight (g/mol) 174.155
MDL Number MFCD00014667
InChI Key FSMYWBQIMDSGQP-UHFFFAOYSA-N
Synonym chromone-3-carboxaldehyde, 3-formylchromone, 4-oxo-4h-chromene-3-carbaldehyde, 3-chromonecarbaldehyde, 4-oxo-4h-1-benzopyran-3-carboxaldehyde, 3-formylchromones, 4h-1-benzopyran-3-carboxaldehyde, 4-oxo, chembl86905, 3-formyl-4-oxo-4h-chromene, chromone-3-carbaldehyde
PubChem CID 87112
IUPAC Name 4-oxochromene-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=O)C(=CO2)C=O

Specifications

Melting Point 150°C to 154°C
Quantity 5 g
Beilstein 472849
Sensitivity Air sensitive
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Chromone-3-carboxaldehyde
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 241-451-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only