missing translation for 'onlineSavingsMsg'
Learn More

Chromomycin A3, 97%

Catalog No. AAJ62927MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62927MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62927MA Supplier Thermo Scientific Chemicals Supplier No. J62927MA
Only null left

CAS: 7059-24-7 | C57H82O26 | 1183.257 g/mol

Chromomycin A3, is used as a G-C specific DNA ligand which inhibits transcription and acts as a DNA binding dye. Chromomycin A3 antagonizes enhanced DNA binding of the transcription factors Sp1 and Sp3 to their cognate G-C box, induced by oxidative stress or DNA damage. Furthermore, by inhibiting transcription, Chromomycin A3 and its structural analogs can inhibit protein biosynthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromomycin A3, is used as a G-C specific DNA ligand which inhibits transcription and acts as a DNA binding dye. Chromomycin A3 antagonizes enhanced DNA binding of the transcription factors Sp1 and Sp3 to their cognate ″G-C&Pr

Chemical Identifiers

CAS 7059-24-7
Molecular Formula C57H82O26
Molecular Weight (g/mol) 1183.257
MDL Number MFCD00043151
InChI Key ZYVSOIYQKUDENJ-YVQLHGKOSA-N
PubChem CID 23928042
IUPAC Name [(2R,3R,4R,6S)-6-[[(6S,7S)-6-[(2S,4R,5R,6R)-4-[(4R,5S,6R)-4-[(2S,4S,5R,6R)-5-acetyloxy-4-hydroxy-4,6-dimethyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-7-[(1S,3S,4R)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-4,10-dihydroxy-3-
SMILES CC1C(C(CC(O1)OC2C(CC3=C(C2=O)C(=C4C(=C3)C=C(C(=C4O)C)OC5CC(C(C(O5)C)OC(=O)C)OC6CC(C(C(O6)C)OC)O)O)C(C(=O)C(C(C)O)O)OC)OC7CC(C(C(O7)C)O)OC8CC(C(C(O8)C)OC(=O)C)(C)O)O

Specifications

Type Chromomycin A3
Physical Form Powder
Melting Point 185°C (decomposition)
Color Yellow
Quantity 10 mg
UN Number UN3462
Merck Index 14,2238
Solubility Information Soluble in ethanol,DMSO,and ethyl acetate (10mg/ml).
Formula Weight 1183.3
Percent Purity 97%
Chemical Name or Material Chromomycin A3
Show More Show Less
Hazard Category H300-H360
Hazard Statement GHS H Statement
H300-H360
Fatal if swallowed.
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
EINECSNumber 230-348-0
RTECSNumber GB7875000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only