missing translation for 'onlineSavingsMsg'
Learn More

Chromium(VI) oxide, 99.5% (metals basis)

Catalog No. p-7048494
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1069114 25 g
AA1069122 100 g
AA1069136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1069114 Supplier Thermo Scientific Chemicals Supplier No. 01069114
Only null left

CAS: 1333-82-0 | CrO3 | 99.99 g/mol

Chromium(VI) oxide is widely used in chromium electroplating, and as protective coatings in corrosion/oxidation resistance of metals. It is also employed in CDs and DVDs. It is useful as an oxidant in Jones oxidation in acetic acid or acetone. It is used in the oxidation of primary alcohols into carboxylic acids, and secondary alcohols into ketones. With phosphoric acid, it is used as a stripping agent for anodic coatings of all types and in the production of syntheric rubies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Boiling Point 250°C (decomposition)
Melting Point 196°C
Color Red
Physical Form Crystalline
Quantity 25 g
UN Number UN1463
Merck Index 14,2235
Solubility Information Very soluble in water. Soluble in sulfuric,nitric acid,acetic acid,and acetone.
Formula Weight 99.99
Percent Purity 99.9%
Sensitivity Hygroscopic
Density 2.7 g/mL
Assay (metals basis)
Chemical Name or Material Chromium(VI) oxide
Show More Show Less
Hazard Category H271-H301-H310+H330-H314-H317-H334-H335-H340-H350-H361f-H372
Hazard Statement GHS H Statement
H271-H301-H310-H330-H334-H340-H350-H372-H361-H314-H318-H317
May cause fire or explosion
strong oxidizer.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P220-P221-P260-P262-P264b-P270-P271-P272-P280g-P281-P283-P284-P285-P302+P350-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P333+P313-P353-P363-P370+P378q-P3
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CHROMIUM TRIOXIDE, ANHYDROUS
EINECSNumber 215-607-8
RTECSNumber GB6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only