missing translation for 'onlineSavingsMsg'
Learn More

Hexavalent Chromium ICP Standard, 1000 ppm Cr6+ in 3% HNO3, Ricca Chemical

Catalog No. PCR61KN500
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PCR61KN500 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PCR61KN500 Supplier Ricca Chemical Company Supplier No. PCR61KN500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 7789-09-5, 7697-37-2, 7732-18-5
Concentration or Composition Notes 1,000ppm Cr
CAS Min % 94.0, 4.22, 0.23
CAS Max % 97.0, 4.39, 0.24
Equivalence 1 mL = 1 mg Cr
Molecular Formula Cr2H8N2O7
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Molecular Weight (g/mol) 252.06
PubChem CID 24600
Chemical Name or Material Chromium(VI) ICP Standard
Show More Show Less