missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium Trioxide, Flakes, Purified, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 125 g |
| Grade | Purified |
| Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.