missing translation for 'onlineSavingsMsg'
Learn More

Chromium powder, -200 mesh, 99.94% (metals basis)

Catalog No. p-4781855
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1379614 25 g
AA1379622 100 g
AA1379636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1379614 Supplier Thermo Scientific Chemicals Supplier No. 01379614
Only null left

Chromium, Cr, CAS Number-7440-47-3, chrome, chromium, elemental, ccris 159, chrome french, compounds, chrom, chrom german, cromo, metal, chromium, metal, 500g, Gray powder, 99.95% (Metals basis), CHEBI:28073, DOT Class: 4.1, Packaging Group: III, 231-157-5, H228 | CAS: 7440-47-3 | Cr | 52.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]

Specifications

Color Silver
Quantity 25 g
Mesh Size −200 mesh
Odor Odorless
UN Number UN3089
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Chromium powder
Hazard Category H228
Hazard Statement GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 231-157-5
RTECSNumber GB4200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only