Learn More
Chromium powder, -10+20 mesh, 99.996% (metals basis)
CAS: 7440-47-3 | Cr | 52.00 g/mol
Supplier: Thermo Scientific Chemicals 01045214
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Gray | |
| 7440-47-3 | |
| 7.19g/cm3 | |
| -10+20 Mesh | |
| 0 hPa | |
| 99.996% (Metals basis) | |
| MFCD00010944 | |
| VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
| chromium | |
| 23976 |
| Chromium | |
| Gray powder | |
| 25 g | |
| Odorless | |
| Powder | |
| Cr | |
| chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
| [Cr] | |
| 52.00 | |
| CHEBI:28073 |
Chemical Identifiers
| 7440-47-3 | |
| 52.00 | |
| VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
| 23976 | |
| chromium |
| Cr | |
| MFCD00010944 | |
| chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
| CHEBI:28073 | |
| [Cr] |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 231-157-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only