missing translation for 'onlineSavingsMsg'
Learn More

Chromium Potassium Sulfate, Dodecahydrate, Technical, Spectrum™ Chemical

Catalog No. p-7165325
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609771 500 g Amber Glass Bottle
18609774 100 lb. Fiber Drum
18609772 2.5 kg Amber Glass Bottle
18609773 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609771 Supplier Spectrum Chemical Mfg Cor Supplier No. C1261500GM
Only null left

Chemical Identifiers

CAS 7788-99-0

Specifications

CAS Max % 1
Quantity 500 g
Grade Technical
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.