missing translation for 'onlineSavingsMsg'
Learn More

Chromium Potassium Sulfate, Dodecahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18609768
Change view
Click to view available options
Quantity:
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609768 500 g Amber Glass Bottle
18609769 2.5 kg Amber Glass Bottle
18609770 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609768 Supplier Spectrum Chemical Mfg Cor Supplier No. C1260500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7788-99-0

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 98 to 102%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.