Learn More
Chromium potassium sulfate dodecahydrate, 98+%
CAS: 7788-99-0 | CrKO8S2·12H2O | 499.39 g/mol
Supplier: Thermo Scientific Chemicals 222525000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Chromium potassium sulfate dodecahydrate | |
| p.a. | |
| Purple | |
| 500 g | |
| CrKO8S2·12H2O | |
| MFCD00149917 | |
| chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p | |
| ZFVHBEKVAITXHW-UHFFFAOYSA-J | |
| potassium;chromium(3+);disulfate;dodecahydrate | |
| 24596 | |
| ≥98% | |
| 1.83 |
| 89.0°C | |
| 7788-99-0 | |
| Crystals | |
| 98.0 to 102% (Iodometry) | |
| CrK(SO4)2·12H2O | |
| 15, 2233 | |
| Solubility in water: 24 g/100 mL (15°C). | |
| O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3] | |
| 499.39 | |
| 499.39 | |
| Plastic bottle | |
| 1.8300g/mL |
Chemical Identifiers
| 7788-99-0 | |
| 499.39 | |
| ZFVHBEKVAITXHW-UHFFFAOYSA-J | |
| 24596 | |
| O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3] |
| CrKO8S2·12H2O | |
| MFCD00149917 | |
| chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p | |
| potassium;chromium(3+);disulfate;dodecahydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
GHS Signal Word: Warning
RTECSNumber : GB6850000
RUO – Research Use Only