missing translation for 'onlineSavingsMsg'
Learn More

Chromium potassium sulfate dodecahydrate, 98+%

CAS: 7788-99-0 | CrKO8S2·12H2O | 499.39 g/mol

Supplier:  Thermo Scientific Chemicals 222521000

 View more versions of this product

Catalog No. AC222521000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chromium potassium sulfate dodecahydrate
p.a.
Purple
100 g
CrKO8S2·12H2O
MFCD00149917
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
ZFVHBEKVAITXHW-UHFFFAOYSA-J
potassium;chromium(3+);disulfate;dodecahydrate
24596
≥98%
1.83
89.0°C
7788-99-0
Crystals
98.0 to 102% (Iodometry)
CrK(SO4)2·12H2O
15, 2233
Solubility in water: 24 g/100 mL (15°C).
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
499.39
499.39
Plastic bottle
1.8300g/mL

Chemical Identifiers

7788-99-0
499.39
ZFVHBEKVAITXHW-UHFFFAOYSA-J
24596
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
CrKO8S2·12H2O
MFCD00149917
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
potassium;chromium(3+);disulfate;dodecahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi

GHS Signal Word: Warning

RTECSNumber : GB6850000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only