missing translation for 'onlineSavingsMsg'
Learn More

Chromium Oxide, Powder, BAKER™, J.T. Baker™

Catalog No. p-5390735
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002602 500 g Poly Bottle
02002601 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002602 Supplier Avantor Supplier No. 161601
Only null left

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
IUPAC Name dichromium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

Physical Form Powder
Quantity 500 g
Formula Weight 151.99
Packaging Poly Bottle
Specific Gravity 1L = 5.10kg
Chemical Name or Material Chromium Oxide, Formerly C.P.