missing translation for 'onlineSavingsMsg'
Learn More

Chromium Metal, 2 in. and Finer, Chips, 99%, Spectrum™ Chemical

Catalog No. p-7165318
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609754 125 g Poly Bottle
18609755 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18609754 Supplier Spectrum Chemical Mfg Cor Supplier No. C1233125GM
Only null left

Chemical Identifiers

CAS 7440-47-3
Molecular Weight (g/mol) 52

Specifications

CAS Max % 1
Quantity 125 g
Packaging Poly Bottle
Form Irregular Pieces
Percent Purity 99%
Grade Ungraded
Chemical Name or Material Chromium Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.