missing translation for 'onlineSavingsMsg'
Learn More

Chromium Metal, 100 Mesh, Powder, 98%, Spectrum™ Chemical

Catalog No. 18609751
Change view
Click to view available options
Quantity:
125 g
500 g
45 kg
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609751 125 g Poly Bottle
18609753 500 g Poly Bottle
18609752 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609751 Supplier Spectrum Chemical Mfg Cor Supplier No. C1234125GM

Chemical Identifiers

CAS 7440-47-3

Specifications

CAS Max % 1
Quantity 125 g
Packaging Poly Bottle
Percent Purity 98%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.