missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) sulfate hydrate, Reagent Grade

Catalog No. aa3330336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3330336 500 g
AA3330322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3330336 Supplier Thermo Scientific Chemicals Supplier No. 03330336

CAS: 15244-38-9 | Cr2O12S3 | 392.16 g/mol

Chromium(III) sulfate hydrate is widely used in making chrome tanned leather, paints, inks and ceramics. It is also used in preparation of chromium metal, dye, in printing and dyeing industry. It is involved in the preparation of chrome catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15244-38-9
Molecular Formula Cr2O12S3
Molecular Weight (g/mol) 392.16
MDL Number MFCD00016039
InChI Key GRWVQDDAKZFPFI-UHFFFAOYSA-H
IUPAC Name dichromium(3+) trisulfate
SMILES [Cr+3].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Purple
Physical Form Crystals
Quantity 500 g
Solubility Information Soluble in water and alcohol. Insoluble in acids.
Formula Weight 392.17 (Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Chromium(III) sulfate hydrate
Hazard Category H302+H332-H315-H317-H319
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 233-253-2
RTECSNumber GB7200000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only