missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) oxide, 99%, amorphous powder

CAS: 1308-38-9 | Cr2O3 | 151.99 g/mol

$131.48 - $131.48

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
IUPAC Name dichromium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC192085000
SDS View Documents
Thermo Scientific Chemicals
192085000
500 g Glass bottle
Each for $131.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1308-38-9
151.99
UOUJSJZBMCDAEU-UHFFFAOYSA-N
517277
dichromium(3+) trioxidandiide
Cr2O3
MFCD00010949
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
CHEBI:48242
[O--].[O--].[O--].[Cr+3].[Cr+3]

Specifications

4000°C
1308-38-9
5 to 7
500 g
Cr2O3
MFCD00010949
15,2238
Solubility in water: insoluble in water. Other solubilities: slightly soluble in acids and alkalies,practically insoluble in alcohol and acetone
[O--].[O--].[O--].[Cr+3].[Cr+3]
151.99
CHEBI:48242
99%
5.21
Chromium(III) oxide
2435°C
Green
Amorphous Powder
98.5% min
Cr2O3
03,38; 13,338
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
UOUJSJZBMCDAEU-UHFFFAOYSA-N
dichromium(3+) trioxidandiide
517277
151.99
Glass bottle
5.2100g/mL
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-160-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only