missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Nitrate Nonahydrate, puriss. p.a., ≥98.0%, Honeywell Fluka™

Catalog No. p-7120324
Change view
Click to view available options
Quantity:
500 g
25 kg
Packaging:
Drum with Inliner
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60045944 500 g Drum with Inliner
60045943 25 kg Drum with Inliner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045944 Supplier Honeywell Chemicals Supplier No. 31313500G
Only null left

Puriss. p.a.,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7789-02-8
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
MDL Number MFCD00149659
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Synonym chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Melting Point 60°C
Quantity 500 g
Linear Formula Cr(NO3)3 · 9H2O
UN Number UN2720
Formula Weight 400.15g/mol
Percent Purity ≥98.0%
Grade puriss. p.a.
Packaging Drum with Inliner
Density 1.89gcm3
Chemical Name or Material Chromium(Iii) Nitrate Nonahydrate
Hazard Statement P210-P280-P305 + P351 + P338-P337 + P313
Precautionary Statement H272-H315-H319
DOTInformation 5.1/PGIII
EINECSNumber 236-921-1
RTECSNumber GB6300000