missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) nitrate nonahydrate, 99.99% min (metals basis)

Catalog No. p-7058754
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4318206 5 g
AA4318214 25 g
AA4318222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4318206 Supplier Thermo Scientific Chemicals Supplier No. 04318206
Only null left

CAS: 7789-02-8 | CrN3O9 | 238.01 g/mol

Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7789-02-8
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
MDL Number MFCD00149659
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
Synonym chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Melting Point 60°C
Color Purple
Physical Form Crystalline
Quantity 5 g
UN Number UN2720
Merck Index 14,2226
Solubility Information Soluble in water and alcohol.
Formula Weight 400.14 (238.01 Anhydrous)
Percent Purity ≥99.99%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Chromium (III) nitrate nonahydrate
Show More Show Less
Hazard Category H272-H317-H319-H332
Hazard Statement GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CHROMIUM NITRATE
EINECSNumber 236-921-1
RTECSNumber GB6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only