missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride hexahydrate, 99.5% min

CAS: 10060-12-5 | Cl3CrH12O6 | 266.44 g/mol

$146.52 - $483.70

Chemical Identifiers

CAS 10060-12-5
Molecular Formula Cl3CrH12O6
Molecular Weight (g/mol) 266.44
MDL Number MFCD00149660
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
PubChem CID 104957
ChEBI CHEBI:53442
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4211422
View Documents
Thermo Scientific Chemicals
04211422
100 g
Each for $146.52
Only null left
 
AA4211436
View Documents
Thermo Scientific Chemicals
04211436
500 g
Each for $483.70
Only null left
 
Description

Description

Chromium(III) chloride hexahydrate is used as a catalyst, as a mordant in printing and dyeing industry. It is involved in the preparation of anhydrous tetrahydrofuran complex of chromium(III) chloride by reacting with trimethylsilylchloride. It finds application in the manufacturing of a variety of chromium-containing pigment in Paint industry and other chromium salts in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10060-12-5
266.44
LJAOOBNHPFKCDR-UHFFFAOYSA-K
104957
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
Cl3CrH12O6
MFCD00149660
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
CHEBI:53442

Specifications

83°C
Black to Green
100 g
MFCD00149660
14,2222
Slightly soluble in acetone. Soluble in water,alcohol. Insoluble in ether
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
266.44
CHEBI:53442
≤99.5%
Hygroscopic
Chromium(III) chloride hexahydrate
10060-12-5
Crystalline
Cl3CrH12O6
UN3260
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
LJAOOBNHPFKCDR-UHFFFAOYSA-K
trichlorochromium;hexahydrate
104957
266.45 (158.35 Anhydrous)
Odorless
1.76 g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H318-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c

H290-H302-H317

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 233-038-3

RTECSNumber : GB5450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only