Learn More
Chromium(III) chloride hexahydrate, 98%
Description
Chromium(III) chloride hexahydrate is used as a catalyst, as a mordant in printing and dyeing industry. It is involved in the preparation of anhydrous tetrahydrofuran complex of chromium(III) chloride by reacting with trimethylsilylchloride. It finds application in the manufacturing of a variety of chromium-containing pigment in Paint industry and other chromium salts in the chemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 83°C |
| Color | Green |
| Physical Form | Crystalline |
| Quantity | 250 g |
| UN Number | UN3260 |
| Merck Index | 14,2222 |
| Solubility Information | Slightly soluble in acetone; Soluble in water, alcohol; Insoluble in ether |
| Formula Weight | 266.45 (158.35 Anhydrous) |
| Percent Purity | 98% |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H290-H302-H317 |
| Hazard Statement | GHS H Statement H334-H314-H318-H302-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. May cause an allergic skin reaction. |
| Precautionary Statement | P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 233-038-3 |
| RTECSNumber | GB5450000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only