missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) acetylacetonate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 212°C |
| Color | Maroon to Purple |
| Boiling Point | 340°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 14.4 to 15.3% (Cr) |
| Packaging | Glass Bottle |
| Linear Formula | (CH3COCH=C(CH3)O)3Cr |
| Quantity | 5 g |
| Solubility Information | Solubility in water: 11g/L (20°C). Other solubilities: 2.20g/100g alcohol (20°C),30.85g/100g benzene (20°C),insoluble in petroleum ether |
| Formula Weight | 349.32 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. If eye irritation persists: Get medical advice/attention. Wear protective gloves/protective clothing/eye protec |
| EINECSNumber | 244-526- |
RUO – Research Use Only