missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) acetate hydroxide, Cr 24%
CAS: 39430-51-8 | Cr3(OH)2(OOCCH3)7
$87.74 - $233.01
Chemical Identifiers
| CAS | 39430-51-8 |
|---|---|
| Molecular Formula | Cr3(OH)2(OOCCH3)7 |
| MDL Number | MFCD00058912 |
Description
Occasionally to dehalogenate organic compounds. used as an oxygen scrubber, polymer industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 39430-51-8 | |
| Cr3(OH)2(OOCCH3)7 | |
| MFCD00058912 |
Specifications
| Blue | |
| Powder | |
| Cr3(OH)2(OOCCH3)7 | |
| Soluble in water. Insoluble in acetone. Soluble in hot water | |
| Odorless |
| 39430-51-8 | |
| 500 g | |
| MFCD00058912 | |
| 603.31 | |
| Chromium(III) acetate hydroxide, Cr 24% |
Safety and Handling
P261-P272-P280g-P302+P352-P333+P313-P363-P501c
H317
EINECSNumber : 254-447-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only