missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) acetate hydroxide, Cr 24%

Catalog No. AA1251836
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1251836 500 g
AA12518A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1251836 Supplier Thermo Scientific Chemicals Supplier No. 01251836
Only null left

CAS: 39430-51-8 | Cr3(OH)2(OOCCH3)7

Occasionally to dehalogenate organic compounds. used as an oxygen scrubber, polymer industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39430-51-8
Molecular Formula Cr3(OH)2(OOCCH3)7
MDL Number MFCD00058912

Specifications

Color Blue
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water. Insoluble in acetone. Soluble in hot water
Formula Weight 603.31
Odor Odorless
Chemical Name or Material Chromium(III) acetate hydroxide, Cr 24%
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 254-447-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only