missing translation for 'onlineSavingsMsg'
Learn More

Chromium cubes, typically 6.35mm (0.25in) square, 99.95% (metals basis)

Catalog No. p-7051949
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4231009 10 g
AA4231018 50 g
AA4231030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4231009 Supplier Thermo Scientific Chemicals Supplier No. 04231009
Only null left

CAS: 7440-47-3 | Cr | 52.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]

Specifications

Quantity 10 g
Odor Odorless
Form Cube
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Chromium cubes, Typically 6.35mm (0.25 in.) square
EINECSNumber 231-157-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only