missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium, 99+%, pure, powder
CAS: 7440-47-3 | Cr | 52.00 g/mol
$409.06 - $409.06
Chemical Identifiers
| CAS | 7440-47-3 |
|---|---|
| Molecular Formula | Cr |
| Molecular Weight (g/mol) | 52.00 |
| MDL Number | MFCD00010944 |
| InChI Key | VYZAMTAEIAYCRO-UHFFFAOYSA-N |
| Synonym | chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 |
| PubChem CID | 23976 |
| ChEBI | CHEBI:28073 |
| IUPAC Name | chromium |
| SMILES | [Cr] |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AC225495000
|
Thermo Scientific Chemicals
225495000 |
500 g |
Each for $409.06
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-47-3 | |
| 52.00 | |
| VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
| 23976 | |
| chromium |
| Cr | |
| MFCD00010944 | |
| chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
| CHEBI:28073 | |
| [Cr] |
Specifications
| 1857.0°C | |
| 2642.0°C | |
| 500 g | |
| Cr | |
| chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
| VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
| chromium | |
| 23976 | |
| 52 | |
| Pure |
| 7440-47-3 | |
| 7.2000g/mL | |
| Cr | |
| MFCD00010944 | |
| Solubility in water: insoluble. | |
| [Cr] | |
| 52.00 | |
| CHEBI:28073 | |
| ≥99% | |
| Chromium |
Safety and Handling
GHS H Statement Very toxic to aquatic life.
GHS P Statement Avoid release to the environment. Collect spillage. Dispose of contents/container to an approved waste disposal plant.
Warning
RUO – Research Use Only