missing translation for 'onlineSavingsMsg'
Learn More

Chromic-Sulfuric Acid Cleaning Solution, Reagents

Catalog No. 01335573
Change view
Click to view available options
Volume (Metric):
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01335573 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335573 Supplier Labchem Inc Supplier No. CC2205002.5D

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chromic-Sulfuric Acid Cleaning Solution, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color-coded No
Form Liquid
pH <1
Sterility Non-sterile
Autoclavable Not Autoclavable
Color Brown/Yellow
Packaging Safety Coated Glass
Quantity 2.5 L
Solubility Completely miscible
Volume (Metric) 2.5 L