missing translation for 'onlineSavingsMsg'
Learn More

Chromeazurol S, Solstice

Catalog No. 6002855
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002855 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002855 Supplier Solstice Supplier No. 7268725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Molecular Weight (g/mol) 605.277
MDL Number MFCD00001615
InChI Key FUIZKNBTOOKONL-UHFFFAOYSA-K
PubChem CID 54736314
IUPAC Name trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]

Specifications

UN Number NONH for all modes of transport
Beilstein 4110655
Quantity 25 g
Formula Weight 605.28g/mol
Chemical Name or Material Chromeazurol S