missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chrome Azurol S, MP Biomedicals™
Specifications
Specifications
| Color | Brown |
| Quantity | 10 g |
| Physical Form | Powder |
| Formula Weight | 605.29 |
| Chemical Name or Material | Chrome Azurol S, Dye content: approx. 65% |
Safety and Handling
| EINECSNumber | 216-787-0 |
| Recommended Storage | Store at Room Temperature (15-30°C). |