missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chrome Azurol S (Mordant Blue 29, CI 43825), Reagents
Specifications
Specifications
| Chemical Name or Material | Chrome Azurol S (Mordant Blue 29, CI 43825) |
| CAS | 1667-99-8 |
| Packaging | Glass Bottle |
| Specific Gravity | 1 |
| Molecular Formula | C23H13Cl2Na3O9S |
| pH Color Changes | Color changes from blue to light purple or orange indicating production of catechol type or hydroxamate type siderophores |
| Synonym | Mordant Blue 29, Color Index 43825 |
| Color | Brown or Green or Black |
| Quantity | 5 g |
| Physical Form | Solid |
| Show More |