missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chromatin Immunoprecipitation (ChIP) Assay Kit

Catalog No. 17295MI
Change view
Click to view available options
Quantity:
22 Assays/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17295MI 22 Assays/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17295MI Supplier MilliporeSigma™ Supplier No. 17295

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Protein A agarose/Salmon Sperm DNA
  • All necessary buffers

Used to enrich for specific regions of chromatin that are immunoprecipitated with a specific antibody for a chromatin protein. Works with all eukaryotic cell types

  • Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions 
  • Supplied buffers are sufficient to generate chromatin from up to five 15 cm plates of cultured cells, each plate providing up to 10 chromatin preparations (varies with cell and assay type)

Specifications

For Use With (Application) 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads
Quantity 22 Assays/Kit