missing translation for 'onlineSavingsMsg'
Learn More

Chroman-8-carboxylic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 31457-16-6
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD12198122
InChI Key LOFOWPRKKPHPDW-UHFFFAOYSA-N
Synonym chroman-8-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-8-carboxylic acid, chromane-8-carboxylic acid
PubChem CID 12701138
IUPAC Name 3,4-dihydro-2H-chromene-8-carboxylic acid
SMILES C1CC2=CC=CC(=C2OC1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC76101CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC76101DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

31457-16-6
178.187
LOFOWPRKKPHPDW-UHFFFAOYSA-N
12701138
C1CC2=CC=CC(=C2OC1)C(=O)O
C10H10O3
MFCD12198122
chroman-8-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-8-carboxylic acid, chromane-8-carboxylic acid
3,4-dihydro-2H-chromene-8-carboxylic acid

Specifications

31457-16-6
100.0
C10H10O3
chroman-8-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-8-carboxylic acid, chromane-8-carboxylic acid
C1CC2=CC=CC(=C2OC1)C(=O)O
178.187
178.19
Chroman-8-carboxylic acid
96.5
97%
MFCD12198122
LOFOWPRKKPHPDW-UHFFFAOYSA-N
3,4-dihydro-2H-chromene-8-carboxylic acid
12701138
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents