missing translation for 'onlineSavingsMsg'
Learn More

Chroman-6-carboxylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC32801DA

Quantity 1g
Packaging Amber Glass Bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. CC32801DA

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Spécifications

Identifiants chimiques

103203-84-5
178.19
IFKANGOXGBPILW-UHFFFAOYSA-N
6504216
OC(=O)C1=CC2=C(OCCC2)C=C1
C10H10O3
MFCD08146590
chroman-6-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-6-carboxylic acid, chromane-6-carboxylic acid, chroman-6-carboxylicacid, 2h-1-benzopyran-6-carboxylicacid, 3,4-dihydro, 6-chromanecarboxylic acid, acmc-1c8ke, 3,4-dihydro-2h-benzopyran-6-carboxylic acid
3,4-dihydro-2H-chromene-6-carboxylic acid

Spécifications

Chroman-6-carboxylic acid
96.5
97%
C10H10O3
MFCD08146590
IFKANGOXGBPILW-UHFFFAOYSA-N
3,4-dihydro-2H-chromene-6-carboxylic acid
6504216
97%
103203-84-5
100.0
Amber Glass Bottle
1g
chroman-6-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-6-carboxylic acid, chromane-6-carboxylic acid, chroman-6-carboxylicacid, 2h-1-benzopyran-6-carboxylicacid, 3,4-dihydro, 6-chromanecarboxylic acid, acmc-1c8ke, 3,4-dihydro-2h-benzopyran-6-carboxylic acid
OC(=O)C1=CC2=C(OCCC2)C=C1
178.19
178.19
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

FDS
Documents

Documents

Promotions

Promotions