Learn More
Chroman-6-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 103203-84-5 |
|---|---|
| Molecular Formula | C10H10O3 |
| Molecular Weight (g/mol) | 178.19 |
| MDL Number | MFCD08146590 |
| InChI Key | IFKANGOXGBPILW-UHFFFAOYSA-N |
| Synonym | chroman-6-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-6-carboxylic acid, chromane-6-carboxylic acid, chroman-6-carboxylicacid, 2h-1-benzopyran-6-carboxylicacid, 3,4-dihydro, 6-chromanecarboxylic acid, acmc-1c8ke, 3,4-dihydro-2h-benzopyran-6-carboxylic acid |
| PubChem CID | 6504216 |
| IUPAC Name | 3,4-dihydro-2H-chromene-6-carboxylic acid |
| SMILES | OC(=O)C1=CC2=C(OCCC2)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
CC32801CB
|
Thermo Scientific Chemicals
CC32801CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC32801DA
|
Thermo Scientific Chemicals
CC32801DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 103203-84-5 | |
| 178.19 | |
| IFKANGOXGBPILW-UHFFFAOYSA-N | |
| 6504216 | |
| OC(=O)C1=CC2=C(OCCC2)C=C1 |
| C10H10O3 | |
| MFCD08146590 | |
| chroman-6-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-6-carboxylic acid, chromane-6-carboxylic acid, chroman-6-carboxylicacid, 2h-1-benzopyran-6-carboxylicacid, 3,4-dihydro, 6-chromanecarboxylic acid, acmc-1c8ke, 3,4-dihydro-2h-benzopyran-6-carboxylic acid | |
| 3,4-dihydro-2H-chromene-6-carboxylic acid |
Specifications
| 103203-84-5 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD08146590 | |
| chroman-6-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-6-carboxylic acid, chromane-6-carboxylic acid, chroman-6-carboxylicacid, 2h-1-benzopyran-6-carboxylicacid, 3,4-dihydro, 6-chromanecarboxylic acid, acmc-1c8ke, 3,4-dihydro-2h-benzopyran-6-carboxylic acid | |
| OC(=O)C1=CC2=C(OCCC2)C=C1 | |
| 178.19 | |
| 178.19 | |
| Chroman-6-carboxylic acid |
| 96.5 | |
| 97% | |
| C10H10O3 | |
| 250mg | |
| IFKANGOXGBPILW-UHFFFAOYSA-N | |
| 3,4-dihydro-2H-chromene-6-carboxylic acid | |
| 6504216 | |
| 97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning