Learn More
Chroman-6-carbonyl chloride, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 55746-03-7 |
|---|---|
| Molecular Formula | C10H9ClO2 |
| Molecular Weight (g/mol) | 196.63 |
| MDL Number | MFCD12198108 |
| InChI Key | DKSNWGZAMLPCLZ-UHFFFAOYSA-N |
| Synonym | chroman-6-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-6-carbonyl chloride, chromane-6-carbonyl chloride, 2h-1-benzopyran-6-carbonyl chloride,3,4-dihydro |
| PubChem CID | 10442680 |
| IUPAC Name | 3,4-dihydro-2H-chromene-6-carbonyl chloride |
| SMILES | C1CC2=C(C=CC(=C2)C(=O)Cl)OC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
CC32802CB
|
Thermo Scientific Chemicals
CC32802CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC32802DA
|
Thermo Scientific Chemicals
CC32802DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 55746-03-7 | |
| 196.63 | |
| DKSNWGZAMLPCLZ-UHFFFAOYSA-N | |
| 10442680 | |
| C1CC2=C(C=CC(=C2)C(=O)Cl)OC1 |
| C10H9ClO2 | |
| MFCD12198108 | |
| chroman-6-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-6-carbonyl chloride, chromane-6-carbonyl chloride, 2h-1-benzopyran-6-carbonyl chloride,3,4-dihydro | |
| 3,4-dihydro-2H-chromene-6-carbonyl chloride |
Specifications
| 55746-03-7 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD12198108 | |
| chroman-6-carbonyl chloride, 3,4-dihydro-2h-1-benzopyran-6-carbonyl chloride, chromane-6-carbonyl chloride, 2h-1-benzopyran-6-carbonyl chloride,3,4-dihydro | |
| C1CC2=C(C=CC(=C2)C(=O)Cl)OC1 | |
| 196.63 | |
| 196.63 | |
| Chroman-6-carbonyl chloride |
| 94.5 | |
| 95% | |
| C10H9ClO2 | |
| 250mg | |
| DKSNWGZAMLPCLZ-UHFFFAOYSA-N | |
| 3,4-dihydro-2H-chromene-6-carbonyl chloride | |
| 10442680 | |
| 95% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger