missing translation for 'onlineSavingsMsg'
Learn More

Chroman-6-carbaldehyde, 95%, Thermo Scientific™

Catalog No. p-3881085
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC32804CB 250mg Amber Glass Bottle
CC32804DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC32804CB Supplier Thermo Scientific Chemicals Supplier No. CC32804CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55745-97-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD08146589
InChI Key YIHDTNNFJDJYRG-UHFFFAOYSA-N
Synonym chroman-6-carbaldehyde, chromane-6-carbaldehyde, 3,4-dihydro-2h-1-benzopyran-6-carbaldehyde, chroman-6-carboxaldehyde, 3,4-dihydro-2h-chromene-6-carboxaldehyde, 2h-1-benzopyran-6-carboxaldehyde, 3,4-dihydro
PubChem CID 6504215
IUPAC Name 3,4-dihydro-2H-chromene-6-carbaldehyde
SMILES C1CC2=C(C=CC(=C2)C=O)OC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material Chroman-6-carbaldehyde
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.