missing translation for 'onlineSavingsMsg'
Learn More

Chroman-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH5844803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5844803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5844803 Supplier Thermo Scientific Chemicals Supplier No. H5844803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 115822-57-6
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD03783554
InChI Key UGAGZMGJJFSKQM-UHFFFAOYSA-N
Synonym chroman-3-carboxylic acid, 3-chromanecarboxylic acid, chromane-3-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-3-carboxylic acid, 2h-1-benzopyran-3-carboxylic acid, 3,4-dihydro, 2h-1-benzopyran-3-carboxylicacid, 3,4-dihydro, 3,4-dihydro-2h-benzopyran-3-carboxylic acid
PubChem CID 2795458
IUPAC Name 3,4-dihydro-2H-chromene-3-carboxylic acid
SMILES C1C(COC2=CC=CC=C21)C(=O)O

Specifications

Melting Point 117°C to 121°C
Quantity 1 g
Formula Weight 178.18
Percent Purity 97%
Chemical Name or Material Chroman-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only