missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences KIMBLE™ CHROMAFLEX™ Flow Adapters

Catalog No. K4208760000
Change view
Click to view available options
Diameter (Metric) Inner:
1 cm
2.5 cm
4.8 cm
Diameter (English) Outer:
0.063 in.
0.125 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Diameter (English) Outer
K4208760000 1 cm 0.063 in.
K4208760020 2.5 cm 0.125 in.
K4208760040 4.8 cm 0.125 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K4208760000 Supplier DWK Life Sciences Supplier No. 4208760000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

KIMBLE™ CHROMAFLEX™ Flow Adapters improve chromatogram resolution by delivering sample and buffer directly to the gel bed top, protecting it from disruption during loading and eliminating dead volume above the gel bed.

  • Flow adapters are recommended when using buffer gradients, during reverse flow applications and for high resolution gravity flow separations
  • Each flow adapter is supplied with a 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 feet (1.5 meters) of FEP PTFE tubing
  • Blue lock ring is supplied with all sizes except those that fit 1.0 cm column ID
  • Use of aggressive organic solvents requires the PTFE bed supports listed as accessories and FFKM o-rings sizes 010, 116 or 222

Specifications

Type Adapter
Diameter (English) Outer 0.063 in.
Diameter (Metric) Inner 1 cm
Temperature (English) Max. 122°F
For Use With (Application) Improving Chromatogram Resolution
Includes 20 micron porosity HDPE bed support, TFE/propylene o-ring seal, stainless steel adjusting rod, PTFE body, and 5 ft. (1.5 meters) of FEP PTFE tubing
Description Fits column ID: 1 cm
For Use With Column, Acrylic Jacketed
Material Polypropylene