Learn More
Chemglass Life Sciences Column, Chromatography, 24/40 Outer Joint, 2in ID X 18in E.L., 2mm Stopcock
Supplier: Chemglass Inc CG118920
Column, Chromatography, 24/40 Outer Joint, 2in ID X 18in E.L., 2mm Stopcock. Similar to CG-1188 but with a sealed in COARSE porosity fritted disc to support column packing. Column is constructed using MEDIUM WALL TUBING. Can be used as a gravity eluted column or a flash column (as described by W.C. Still, M. Kahn and A. Mitra of Columbia University in JOURNAL OF ORGANIC CHEMISTRY 43, 2923 (1978), and covered in U.S. Patent Number 4,293,422 dated October 6, 1981) when used with optional CG-1191 flow controller and CG-1190 reservoir.