missing translation for 'onlineSavingsMsg'
Learn More

Choline Chloride 98.0+%, TCI America™

Catalog No. p-7124358
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
C032925G 25 g
C0329500G 500 g
2 missing translation for 'options'

Catalog No. C032925G

Supplier: TCI America C032925G

Only null left

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point 302°C
Color White
Quantity 25 g
Formula Weight 139.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Choline Chloride
EINECSNumber (9)-1994&(2)-0341
RTECSNumber KH2975000
TSCA Yes