missing translation for 'onlineSavingsMsg'
Learn More

Choline Chloride 98.0+%, TCI America™

Catalog No. C032925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C032925G 25 g
C0329500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C032925G Supplier TCI America Supplier No. C032925G

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point 302°C
Color White
Quantity 25 g
Formula Weight 139.62
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Choline Chloride
EINECSNumber (9)-1994&(2)-0341
RTECSNumber KH2975000
TSCA Yes