missing translation for 'onlineSavingsMsg'
Learn More

Choline Chloride Dietary Supplement, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. 18601678
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601678 500 g Poly Bottle
18601679 2.5 kg Poly Pail
18601680 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18601678 Supplier Spectrum Chemical Mfg Cor Supplier No. C2058500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

C2058, 67-48-1, C5H14ClNO

Spectrum™ Chemical Choline Chloride, USP Dietary Supplement is an essential nutrient and typically grouped with B-complex vitamins. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
IUPAC Name (2-hydroxyethyl)trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 99 to 100.5%
Grade USP Dietary Supplement
Ignition Residue 0.05%
Packaging Poly Bottle