missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline chloride, 98+%
Description
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
Solubility
Soluble in water, ethanol, methanol and alcohol.
Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
Choline chloride helps in providing nourishment for pig feed and poultry feed. It acts as a food flavor enhancer. It is used as a precursor of acetylcholine and is a methyl donor in various metabolic processes and in lipid metabolism.
Solubility
Soluble in water, ethanol, methanol and alcohol.
Notes
Incompatible with strong oxidizing agents, strong bases, strong acids, alkalies and alloys.
Specifications
Specifications
| Melting Point | ∽305°C (decomposition) |
| Assay Percent Range | ≥98% |
| Quantity | 100 g |
| Beilstein | 3563126 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,2206 |
| Solubility Information | Soluble in water,ethanol,methanol and alcohol. |
| Formula Weight | 139.63 |
| Percent Purity | ≥98% |
| Physical Form | Solid |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a |
| EINECSNumber | 200-655-4 |
| RTECSNumber | KH2975000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only