missing translation for 'onlineSavingsMsg'
Learn More

Choline chloride, 98+% (dry basis)

Catalog No. AAJ13410P5
Change view
Click to view available options
Quantity:
5 lb.
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ13410P5 5 lb. Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ13410P5 Supplier Thermo Fisher Scientific Supplier No. J13410P5
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 67-48-1 | C5H14ClNO | 139.62 g/mol

Chemical Identifiers

CAS 67-48-1
Molecular Formula C5H14ClNO
Molecular Weight (g/mol) 139.62
MDL Number MFCD00011721
InChI Key SGMZJAMFUVOLNK-UHFFFAOYSA-M
Synonym choline chloride, hepacholine, biocolina, biocoline, lipotril, paresan, luridin chloride, choline hydrochloride, cholinium chloride, bilineurin chloride
PubChem CID 6209
ChEBI CHEBI:133341
SMILES [Cl-].C[N+](C)(C)CCO

Specifications

Melting Point ∼305°C (decomposition)
Packaging Plastic bottle
Quantity 5 lb.
Beilstein 3563126
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;chloride
Formula Weight 139.62
Percent Purity ≥98%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material Choline chloride
EINECSNumber 200-655-4
RTECSNumber KH2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only