missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Bromide 98.0+%, TCI America™
$140.54 - $1205.16
Chemical Identifiers
| CAS | 1927-06-6 |
|---|---|
| Molecular Formula | C5H14BrNO |
| Molecular Weight (g/mol) | 184.08 |
| MDL Number | MFCD00054244 |
| InChI Key | JJCWKVUUIFLXNZ-UHFFFAOYSA-M |
| Synonym | (2-Hydroxyethyl)trimethylammonium Bromide |
| PubChem CID | 74724 |
| IUPAC Name | 2-hydroxyethyl(trimethyl)azanium;bromide |
| SMILES | C[N+](C)(C)CCO.[Br-] |
Chemical Identifiers
| 1927-06-6 | |
| 184.08 | |
| JJCWKVUUIFLXNZ-UHFFFAOYSA-M | |
| 74724 | |
| C[N+](C)(C)CCO.[Br-] |
| C5H14BrNO | |
| MFCD00054244 | |
| (2-Hydroxyethyl)trimethylammonium Bromide | |
| 2-hydroxyethyl(trimethyl)azanium;bromide |
Specifications
| 1927-06-6 | |
| White | |
| MFCD00054244 | |
| (2-Hydroxyethyl)trimethylammonium Bromide | |
| C[N+](C)(C)CCO.[Br-] | |
| 184.08 | |
| 184.08 | |
| Crystalline Powder |
| 298°C | |
| C5H14BrNO | |
| 25 g | |
| JJCWKVUUIFLXNZ-UHFFFAOYSA-M | |
| 2-hydroxyethyl(trimethyl)azanium;bromide | |
| 74724 | |
| ≥98.0% (T) | |
| Choline Bromide |
Safety and Handling
TSCA : No