missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Choline Bitartrate Dietary Supplement, USP, 99-100.5%, Spectrum™ Chemical
C2052, 87-67-2, C9H19NO7
Supplier: Spectrum Chemical Mfg Cor C205212KG
Description
Spectrum™ Chemical Choline Bitartrate, USP Dietary Supplement is an essential nutrient and typically grouped with B-complex vitamins. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 87-67-2 | |
| 0.1% | |
| C9H19NO7 | |
| QWJSAWXRUVVRLH-LREBCSMRSA-M | |
| C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O | |
| 253.25 | |
| USP Dietary Supplement |
| 100% | |
| Poly Pail | |
| 12 kg | |
| +17.5 to +18.5° | |
| (2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate | |
| 99-100.5% |
Chemical Identifiers
| 87-67-2 | |
| 253.25 | |
| (2-hydroxyethyl)trimethylazanium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate |
| C9H19NO7 | |
| QWJSAWXRUVVRLH-LREBCSMRSA-M | |
| C[N+](C)(C)CCO.O[C@H]([C@@H](O)C([O-])=O)C(O)=O |